Learn More
Grainger Cut-Off TKF 12L050S PR1535 10 pcs.
Catalog No. 502764801
Encompass_Preferred
The Grainger Cut-Off TKF 12L050S PR1535 is a 10-pack of high-speed steel cutting tools with a 12mm shank diameter and 50mm cutting diameter. They have a right hand spiral flute design and are designed for use on lathes, automatic lathes, and screw machines. The tools feature TiCN coating for wear resistance and have an ISO P15 blade configuration with 3 flutes. Overall length is 125mm.
Catalog No. 50-276-4801 Supplier Grainger Supplier No. 170FY3
The Grainger Cut-Off TKF 12L050S PR1535 is a 10-pack of high-speed steel cutting tools with a 12mm shank diameter and 50mm cutting diameter. They have a right hand spiral flute design and are designed for use on lathes, automatic lathes, and screw machines. The tools feature TiCN coating for wear resistance and have an ISO P15 blade configuration with 3 flutes. Overall length is 125mm.