Learn More
Grainger Cut-Off TKF 12L050S16DR PR1535 G 10 pcs.
Catalog No. 502764838
Encompass_Preferred
The Grainger 170FY2 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical seal. It has a 115V motor with a 1.25" NPT suction and 1" NPT discharge. This centrifugal pump has a max flow rate of 160 GPM, max head of 28 feet, and max pressure of 12 PSI. The pump end and motor are close coupled with a NEMA 56J frame. This is a single stage centrifugal pump ideal for transferring water or light chemicals.
Catalog No. 50-276-4838 Supplier Grainger Supplier No. 170FY2
The Grainger 170FY2 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical seal. It has a 115V motor with a 1.25" NPT suction and 1" NPT discharge. This centrifugal pump has a max flow rate of 160 GPM, max head of 28 feet, and max pressure of 12 PSI. The pump end and motor are close coupled with a NEMA 56J frame. This is a single stage centrifugal pump ideal for transferring water or light chemicals.