Learn More
Grainger Cut-Off TKF 12R050S KW10 U 10 pcs.
Catalog No. 502765793
Encompass_Preferred
The 170EW4 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof Fan Motor with a 1.15 Service Factor from Grainger. It features sleeve bearings and is totally enclosed for protection against dirt, dust, water and other contaminants. This single phase motor can be wired for 115V or 230V operation. It has Class B insulation and a NEMA 48Y frame. The 170EW4 would be a good choice for industrial ventilation, cooling, fume exhaust or material handling applications.
Catalog No. 50-276-5793 Supplier Grainger Supplier No. 170EW4
Add to Cart
Add to Cart
The 170EW4 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof Fan Motor with a 1.15 Service Factor from Grainger. It features sleeve bearings and is totally enclosed for protection against dirt, dust, water and other contaminants. This single phase motor can be wired for 115V or 230V operation. It has Class B insulation and a NEMA 48Y frame. The 170EW4 would be a good choice for industrial ventilation, cooling, fume exhaust or material handling applications.