Learn More
Grainger Cut-Off TKF 16L150S PR1225 10 pcs.
Catalog No. 502766296
Encompass_Preferred
The Grainger 170FG1 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings, manual reset thermal overload protection, and a cast iron body. The motor is CSA certified and can be used for driving pumps, fans, and other machinery. With its 1/3 horsepower, it provides good low to medium power performance for light industrial and commercial applications. The motor dimensions are 6-1/2" diameter x 8-3/16" long and it weighs 15 pounds.
Catalog No. 50-276-6296 Supplier Grainger Supplier No. 170FG1
The Grainger 170FG1 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings, manual reset thermal overload protection, and a cast iron body. The motor is CSA certified and can be used for driving pumps, fans, and other machinery. With its 1/3 horsepower, it provides good low to medium power performance for light industrial and commercial applications. The motor dimensions are 6-1/2" diameter x 8-3/16" long and it weighs 15 pounds.