Learn More
Grainger Cut-Off TKF 16L150T16DR PR1535 G 10 pcs.
Catalog No. 502763890
Encompass_Preferred
The 170GD3 is a Grainger Approved 3-Phase AC Induction Motor made by Baldor Electric Company. It has a horsepower of 1, operates on 230/460V at 60Hz, is rated for continuous duty, has a TEFC enclosure, and is C-face mount. The motor is inverter rated, has class F insulation, and 1.15 service factor. It has a NEMA design B torque characteristic and features regreasable bearings. The overall dimensions are 9-7/16" Diameter x 13-3/8" Length.
Catalog No. 50-276-3890 Supplier Grainger Supplier No. 170GD3
Add to Cart
Add to Cart
The 170GD3 is a Grainger Approved 3-Phase AC Induction Motor made by Baldor Electric Company. It has a horsepower of 1, operates on 230/460V at 60Hz, is rated for continuous duty, has a TEFC enclosure, and is C-face mount. The motor is inverter rated, has class F insulation, and 1.15 service factor. It has a NEMA design B torque characteristic and features regreasable bearings. The overall dimensions are 9-7/16" Diameter x 13-3/8" Length.