Learn More
Grainger Cut-Off TKF 16R150S PR1225 10 pcs.
Catalog No. 502765479
Encompass_Preferred
The Grainger 170FG7 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features permanent split capacitor for high starting torque. This electric motor can be used for driving pumps, fans, conveyors, and other shop equipment. It has ball bearings for smooth, reliable operation and a thermally protected design. The overall dimensions are 6-1/2" diameter x 7-7/16" long.
Catalog No. 50-276-5479 Supplier Grainger Supplier No. 170FG7
The Grainger 170FG7 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features permanent split capacitor for high starting torque. This electric motor can be used for driving pumps, fans, conveyors, and other shop equipment. It has ball bearings for smooth, reliable operation and a thermally protected design. The overall dimensions are 6-1/2" diameter x 7-7/16" long.