Learn More
Grainger Cut-Off TKF 16R150T16DR PR1535 G 10 pcs.
Catalog No. 502764365
Encompass_Preferred
The Grainger 170GE3 is a General Electric 3-Phase AC Induction Motor with a horsepower of 1, an RPM of 1725, and a voltage of 230/460V. It has a TEFC enclosure type, operates on 60 Hz power, and has a frame size of 184T. The motor is CSA certified and has a full load amp rating of 3.6/1.8A. It can provide efficient and reliable power for industrial applications requiring a 1 HP motor with a 1725 RPM operating speed.
Catalog No. 50-276-4365 Supplier Grainger Supplier No. 170GE3
The Grainger 170GE3 is a General Electric 3-Phase AC Induction Motor with a horsepower of 1, an RPM of 1725, and a voltage of 230/460V. It has a TEFC enclosure type, operates on 60 Hz power, and has a frame size of 184T. The motor is CSA certified and has a full load amp rating of 3.6/1.8A. It can provide efficient and reliable power for industrial applications requiring a 1 HP motor with a 1725 RPM operating speed.