Learn More
Grainger Cut-Off TKN 16 CR9025 CVD 10 pcs.
Catalog No. 502765443
Encompass_Preferred
The 170ER6 is a 6 volt, 170 amp hour sealed lead acid battery from EnerSys. It is a deep cycle battery designed for solar and other renewable energy applications. The battery measures 10.24" x 7.13" x 10.83" (LxWxH) and weighs 63 pounds. It has threaded terminals for secure connections and can be installed upright or on its side. The battery is maintenance free and has an estimated service life of 7-10 years under proper conditions. With its deep cycle design, the 170ER6 is well-suited for repeated discharges and recharges as required in renewable energy systems.
Catalog No. 50-276-5443 Supplier Grainger Supplier No. 170ER6
The 170ER6 is a 6 volt, 170 amp hour sealed lead acid battery from EnerSys. It is a deep cycle battery designed for solar and other renewable energy applications. The battery measures 10.24" x 7.13" x 10.83" (LxWxH) and weighs 63 pounds. It has threaded terminals for secure connections and can be installed upright or on its side. The battery is maintenance free and has an estimated service life of 7-10 years under proper conditions. With its deep cycle design, the 170ER6 is well-suited for repeated discharges and recharges as required in renewable energy systems.