Learn More
Grainger Cut-Off TKN 2 KW10 Uncoate 10 pcs.
Catalog No. 502766553
Encompass_Preferred
The Grainger 170ET2 is a Dayton 1/3 HP Electric Motor with a totally enclosed fan cooled design. It has a single phase, 115V AC power supply and operates at 3450 RPM. The motor has a 48Y frame with a 1.15 service factor. It can output 0.25 horsepower and has manual overload protection. The motor dimensions are 6-1/2" diameter x 8-3/16" long. It has a C-face mounting style and 1/2" diameter shaft. The motor is thermally protected and suitable for OEM and replacement applications.
Catalog No. 50-276-6553 Supplier Grainger Supplier No. 170ET2
The Grainger 170ET2 is a Dayton 1/3 HP Electric Motor with a totally enclosed fan cooled design. It has a single phase, 115V AC power supply and operates at 3450 RPM. The motor has a 48Y frame with a 1.15 service factor. It can output 0.25 horsepower and has manual overload protection. The motor dimensions are 6-1/2" diameter x 8-3/16" long. It has a C-face mounting style and 1/2" diameter shaft. The motor is thermally protected and suitable for OEM and replacement applications.