Learn More
Grainger Cut-Off TKN 24 TN90 Uncoat 10 pcs.
Catalog No. 502765432
Encompass_Preferred
The Grainger 170ET0 is a 3/4 HP, 115V, single phase, open drip proof electric motor with a 1075 RPM base speed and a 1.15 service factor. It has a 48 frame size, features sleeve bearings, and includes a conduit box. The motor is thermally protected and rated for continuous duty applications.
Catalog No. 50-276-5432 Supplier Grainger Supplier No. 170ET0
The Grainger 170ET0 is a 3/4 HP, 115V, single phase, open drip proof electric motor with a 1075 RPM base speed and a 1.15 service factor. It has a 48 frame size, features sleeve bearings, and includes a conduit box. The motor is thermally protected and rated for continuous duty applications.