Learn More
Grainger Cut-Off TKN 2P TN90 Uncoat 10 pcs.
Catalog No. 502766331
Encompass_Preferred
The Grainger 170EV5 is a 5-gallon plastic pail with lid that is ideal for storing and transporting a variety of items. It is made from durable high density polyethylene with a bail handle for easy carrying. The snap-on lid helps keep contents secure. This pail is chemical resistant and FDA approved for food storage. It measures 12" diameter x 13" high and has a capacity of 5 gallons. The pail is white in color. With its sturdy construction and sealable lid, this pail is great for commercial and industrial use.
Catalog No. 50-276-6331 Supplier Grainger Supplier No. 170EV5
The Grainger 170EV5 is a 5-gallon plastic pail with lid that is ideal for storing and transporting a variety of items. It is made from durable high density polyethylene with a bail handle for easy carrying. The snap-on lid helps keep contents secure. This pail is chemical resistant and FDA approved for food storage. It measures 12" diameter x 13" high and has a capacity of 5 gallons. The pail is white in color. With its sturdy construction and sealable lid, this pail is great for commercial and industrial use.