Learn More
Grainger Cut-Off TKN 3 CR9025 CVD C 10 pcs.
Catalog No. 502765779
Encompass_Preferred
The 170ET3 is a 3-phase, 60 Hz, 460 V AC, 3 HP, 1725 RPM, TEFC electric motor made by Baldor-Reliance. It has a cast iron frame, thermally protected, and is C-face mount with a 182T frame. The motor is designed for V-belt drives, has a 1.15 service factor, and includes a conduit box. The overall dimensions are 9-7/16" diameter x 15-3/16" long. This general purpose motor can be used for industrial machinery requiring 3 HP.
Catalog No. 50-276-5779 Supplier Grainger Supplier No. 170ET3
The 170ET3 is a 3-phase, 60 Hz, 460 V AC, 3 HP, 1725 RPM, TEFC electric motor made by Baldor-Reliance. It has a cast iron frame, thermally protected, and is C-face mount with a 182T frame. The motor is designed for V-belt drives, has a 1.15 service factor, and includes a conduit box. The overall dimensions are 9-7/16" diameter x 15-3/16" long. This general purpose motor can be used for industrial machinery requiring 3 HP.