Learn More
Grainger Cut-Off TKN 3 TN90 Uncoate 10 pcs.
Catalog No. 502766332
Encompass_Preferred
The Grainger 170ET5 is a 5-gallon plastic utility pail made in the USA by Encore Plastics. It features a tapered body for easy nesting and stacking, a reinforced bottom for durability, a bail handle for easy carrying, and graduation marks up to 5 gallons for liquid measurement. The pail is constructed from high density polyethylene and has a maximum temperature rating of 120°F. It meets FDA regulations for food contact. The pail has an approximate 9" top opening and is 12" high with a 10.5" bottom diameter. It weighs 1.1 lbs and has a capacity of 5 gallons. The pail comes in a natural white color.
Catalog No. 50-276-6332 Supplier Grainger Supplier No. 170ET5
The Grainger 170ET5 is a 5-gallon plastic utility pail made in the USA by Encore Plastics. It features a tapered body for easy nesting and stacking, a reinforced bottom for durability, a bail handle for easy carrying, and graduation marks up to 5 gallons for liquid measurement. The pail is constructed from high density polyethylene and has a maximum temperature rating of 120°F. It meets FDA regulations for food contact. The pail has an approximate 9" top opening and is 12" high with a 10.5" bottom diameter. It weighs 1.1 lbs and has a capacity of 5 gallons. The pail comes in a natural white color.