Learn More
Grainger Cut-Off TKN 4 KW10 Uncoate 10 pcs.
Catalog No. 502765495
Encompass_Preferred
The 170ET8 is a 3M Scotch-Brite High Performance Industrial Hand Pad. It is a 6 inch by 9 inch hand pad made of high-quality nylon fibers in an open textured nonwoven construction. This pad is designed for industrial cleaning, finishing and deburring applications. It effectively removes rust, scale, paint, dirt and burrs from metal, plastic and fiberglass surfaces. The pad has an open coat abrasive that cuts fast without scratching. It is durable, resists loading and has high scrubbing power. The 3M Scotch-Brite High Performance Industrial Hand Pad 170ET8 provides versatile cleaning and finishing for industrial applications.
Catalog No. 50-276-5495 Supplier Grainger Supplier No. 170ET8
The 170ET8 is a 3M Scotch-Brite High Performance Industrial Hand Pad. It is a 6 inch by 9 inch hand pad made of high-quality nylon fibers in an open textured nonwoven construction. This pad is designed for industrial cleaning, finishing and deburring applications. It effectively removes rust, scale, paint, dirt and burrs from metal, plastic and fiberglass surfaces. The pad has an open coat abrasive that cuts fast without scratching. It is durable, resists loading and has high scrubbing power. The 3M Scotch-Brite High Performance Industrial Hand Pad 170ET8 provides versatile cleaning and finishing for industrial applications.