Learn More
Grainger Cut-Off TKN 4 PR1535 PVD C 10 pcs.
Catalog No. 502764762
Encompass_Preferred
The Grainger 170FX6 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical shaft seal. It has a 115V motor with a 1-1/4" NPT suction and 1" NPT discharge. This centrifugal pump can deliver up to 22 GPM at 15' of head and is designed for clear water applications. The pump end materials make it suitable for water, oil, and mild chemical applications. With its compact size and quiet operation, this versatile pump can be used for transferring, circulating, boosting, spraying, and sampling liquids.
Catalog No. 50-276-4762 Supplier Grainger Supplier No. 170FX6
The Grainger 170FX6 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical shaft seal. It has a 115V motor with a 1-1/4" NPT suction and 1" NPT discharge. This centrifugal pump can deliver up to 22 GPM at 15' of head and is designed for clear water applications. The pump end materials make it suitable for water, oil, and mild chemical applications. With its compact size and quiet operation, this versatile pump can be used for transferring, circulating, boosting, spraying, and sampling liquids.