Learn More
Grainger Cut-Off TKN 48 CR9025 CVD 10 pcs.
Catalog No. 502765476
Encompass_Preferred
The Grainger 170ET6 is a Westinghouse 6-inch LED Recessed Retrofit Downlight. It has a color temperature of 4000K and delivers 800 lumens. The downlight has an adjustable beam spread of 25-45 degrees. It is dimmable and has a CRI of 80. The downlight is designed for use in new construction and remodel applications. It is UL listed for damp locations and has an estimated life of 50,000 hours. The downlight requires an opening of 6.75 inches and has a depth of 5.25 inches. It comes with a 5 year warranty.
Catalog No. 50-276-5476 Supplier Grainger Supplier No. 170ET6
The Grainger 170ET6 is a Westinghouse 6-inch LED Recessed Retrofit Downlight. It has a color temperature of 4000K and delivers 800 lumens. The downlight has an adjustable beam spread of 25-45 degrees. It is dimmable and has a CRI of 80. The downlight is designed for use in new construction and remodel applications. It is UL listed for damp locations and has an estimated life of 50,000 hours. The downlight requires an opening of 6.75 inches and has a depth of 5.25 inches. It comes with a 5 year warranty.