Learn More
Grainger Cut-Off TKN 5 KW10 Uncoate 10 pcs.
Catalog No. 502765792
Encompass_Preferred
The Grainger 170EU0 is a Westinghouse 15 Amp, 125 Volt, 2-Pole, 3-Wire Grounding Plug with a NEMA 5-15P Configuration. It has side wired screw terminals and is UL Listed with a maximum temperature rating of 167°F. This plug is ideal for connecting appliances, power tools, and lighting fixtures to extension cords. The plug's durable thermoplastic construction provides a sturdy connection.
Catalog No. 50-276-5792 Supplier Grainger Supplier No. 170EU0
The Grainger 170EU0 is a Westinghouse 15 Amp, 125 Volt, 2-Pole, 3-Wire Grounding Plug with a NEMA 5-15P Configuration. It has side wired screw terminals and is UL Listed with a maximum temperature rating of 167°F. This plug is ideal for connecting appliances, power tools, and lighting fixtures to extension cords. The plug's durable thermoplastic construction provides a sturdy connection.