Learn More
Grainger Cut-Off TKN 6 CR9025 CVD C 10 pcs.
Catalog No. 502765321
Encompass_Preferred
The 170EV0 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is a stored pressure type extinguisher containing 5 lbs of Novec 1230 fluid. This agent is intended for use on Class A, B and C fires. The extinguisher has a discharge range of 10-15 ft. It features an all-steel valve construction and discharge hose. The extinguisher meets UL and ULC standards and comes with a 5 year limited warranty from the manufacturer. It can be recharged after use. This is an ideal clean agent extinguisher when a non-conductive agent is needed.
Catalog No. 50-276-5321 Supplier Grainger Supplier No. 170EV0
The 170EV0 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is a stored pressure type extinguisher containing 5 lbs of Novec 1230 fluid. This agent is intended for use on Class A, B and C fires. The extinguisher has a discharge range of 10-15 ft. It features an all-steel valve construction and discharge hose. The extinguisher meets UL and ULC standards and comes with a 5 year limited warranty from the manufacturer. It can be recharged after use. This is an ideal clean agent extinguisher when a non-conductive agent is needed.