Learn More
MSC Ansell Gloves, Cut A4, Puncture 3, Abrasion 4, Foam Nitrile Coated
Catalog No. 502835508
Encompass
The gloves are lightweight, industrial work gloves engineered for superior cut, puncture, and abrasion resistance. They combine Kevlar, Nylon, Spandex, and Stainless Steel materials with a foam nitrile-coated palm to deliver excellent grip and protection in demanding environments.
  • Superior cut, puncture, and abrasion resistance
  • Constructed with Kevlar, nylon, spandex, and stainless steel for durability
  • Foam nitrile-coated palm provides enhanced grip
  • Lightweight design ensures comfort during prolonged use
  • Meets ANSI/ISEA A4 cut resistance, 3 puncture resistance, and 4 abrasion resistance
Catalog No. 50-283-5508 Supplier MSC Supplier No. 67008037
The gloves are lightweight, industrial work gloves engineered for superior cut, puncture, and abrasion resistance. They combine Kevlar, Nylon, Spandex, and Stainless Steel materials with a foam nitrile-coated palm to deliver excellent grip and protection in demanding environments.
  • Superior cut, puncture, and abrasion resistance
  • Constructed with Kevlar, nylon, spandex, and stainless steel for durability
  • Foam nitrile-coated palm provides enhanced grip
  • Lightweight design ensures comfort during prolonged use
  • Meets ANSI/ISEA A4 cut resistance, 3 puncture resistance, and 4 abrasion resistance