Learn More
Grainger Superior Glove Dexterity Knit Safety Gloves, Pu Palm Dipped, ASTM Cut 4, Size L (9), 1 Pair
Catalog No. 502783153
Encompass_Preferred
These palm dipped gloves offer reliable protection for a variety of general purpose tasks. Designed with a high-cut resistance level, they provide enhanced safety in environments where handling sharp objects is common.
  • Provides ASTM cut level 4 protection
  • Palm dipped coating for improved grip and dexterity
  • Comfortable knit construction
  • Size large (9) for a secure fit
Catalog No. 50-278-3153 Supplier Grainger Supplier No. 19NE08
These palm dipped gloves offer reliable protection for a variety of general purpose tasks. Designed with a high-cut resistance level, they provide enhanced safety in environments where handling sharp objects is common.
  • Provides ASTM cut level 4 protection
  • Palm dipped coating for improved grip and dexterity
  • Comfortable knit construction
  • Size large (9) for a secure fit