Learn More
Grainger Superior Glove Dexterity Polyurethane Palm Dipped Work Glove, ASTM Cut Level A4, 3XL, 12/Pk.
Catalog No. 502779779
Encompass_Preferred
These 13-gauge work gloves are designed for tasks requiring reliable cut protection and a secure grip. Constructed with an engineered yarn liner, they offer 360° ASTM ANSI A4 cut resistance. The non-marring polyurethane palm coating provides enhanced dry grip and is formulated to ensure no residue is left on handled surfaces.
  • Provides 360° moderate ANSI A4 cut resistance
  • Features non-marring polyurethane palm coating for improved dry grip
  • Designed with an ergonomic hand form for a natural and comfortable fit
  • Maintains good breathability through the uncoated back of hand
  • Rated for ASTM ANSI abrasion resistance level 4 and puncture resistance level 2
  • Wash in hot water and tumble dry with high heat; do not use bleach
Catalog No. 50-277-9779 Supplier Grainger Supplier No. 19NE11
These 13-gauge work gloves are designed for tasks requiring reliable cut protection and a secure grip. Constructed with an engineered yarn liner, they offer 360° ASTM ANSI A4 cut resistance. The non-marring polyurethane palm coating provides enhanced dry grip and is formulated to ensure no residue is left on handled surfaces.
  • Provides 360° moderate ANSI A4 cut resistance
  • Features non-marring polyurethane palm coating for improved dry grip
  • Designed with an ergonomic hand form for a natural and comfortable fit
  • Maintains good breathability through the uncoated back of hand
  • Rated for ASTM ANSI abrasion resistance level 4 and puncture resistance level 2
  • Wash in hot water and tumble dry with high heat; do not use bleach