Learn More
Grainger Cut-Resistant Gloves, L, Palm, Nitrile Glove Coating Material, 1 PR
Catalog No. NC1604567
Encompass_Preferred
These Condor cut-resistant gloves are designed for protection, comfort, and performance in a wide variety of demanding jobs. The foam nitrile coating helps protect against petroleum, acids, and solvents, and resists snags, punctures, abrasion, and cuts. Crinkle finish adds texture for better grip and productivity in tasks from parts handling and automotive assembly to construction, food industries, and metal fabrication. The lightweight, HPPE (high-performance polyethylene) lining offers cut, puncture, and abrasion resistance when handling ceramics, glass, and plastics and in carpentry and joining tasks.
This item is not returnable. View return policy
Catalog No. NC1604567 Supplier Grainger Supplier No. 48UR04
Add to Cart
Add to Cart
These Condor cut-resistant gloves are designed for protection, comfort, and performance in a wide variety of demanding jobs. The foam nitrile coating helps protect against petroleum, acids, and solvents, and resists snags, punctures, abrasion, and cuts. Crinkle finish adds texture for better grip and productivity in tasks from parts handling and automotive assembly to construction, food industries, and metal fabrication. The lightweight, HPPE (high-performance polyethylene) lining offers cut, puncture, and abrasion resistance when handling ceramics, glass, and plastics and in carpentry and joining tasks.