Learn More
Electron Microscopy Sciences Safeknit™ Cut-Resistant Gloves Heavy Duty, Size 9
SDP
Catalog No. 50930744
Encompass_Preferred
High performance cut resistant Safeknit glove is a blend of abrasion-resistant Spectra fiber and other synthetics. The mix of these fibers form a rugged cut resistant barrier that offers the histologist sensitivity for manipulation but with extreme comfort and productivity.
NOTE: These gloves are neither puncture proof nor water resistant. They should not be used to protect against intentional cutting. Avoid use near moving blades. Two strand seamless glove.
Size: 9
This item is not returnable. View return policy
Catalog No. 50-930-744 Supplier Electron Microscopy Sciences Supplier No. 7106509
High performance cut resistant Safeknit glove is a blend of abrasion-resistant Spectra fiber and other synthetics. The mix of these fibers form a rugged cut resistant barrier that offers the histologist sensitivity for manipulation but with extreme comfort and productivity.
NOTE: These gloves are neither puncture proof nor water resistant. They should not be used to protect against intentional cutting. Avoid use near moving blades. Two strand seamless glove.
Size: 9