Learn More
Electron Microscopy Sciences Safeknit™ Cut-Resistant Gloves Medium Duty, Size 6
SDP
Catalog No. 501899451
Encompass_Preferred
High performance cut resistant Safeknit glove is a blend of abrasion-resistant Spectra fiber and other synthetics. The mix of these fibers form a rugged cut resistant barrier that offers the histologist sensitivity for manipulation but with extreme comfort and productivity.
NOTE: These gloves are neither puncture proof nor water resistant. They should not be used to protect against intentional cutting. Avoid use near moving blades. Single strand seamless glove.
Size: 6
This item is not returnable. View return policy
Catalog No. 50-189-9451 Supplier Electron Microscopy Sciences Supplier No. 7106606
High performance cut resistant Safeknit glove is a blend of abrasion-resistant Spectra fiber and other synthetics. The mix of these fibers form a rugged cut resistant barrier that offers the histologist sensitivity for manipulation but with extreme comfort and productivity.
NOTE: These gloves are neither puncture proof nor water resistant. They should not be used to protect against intentional cutting. Avoid use near moving blades. Single strand seamless glove.
Size: 6