Learn More
Active Motif CUT & Tag-IT Assay Kit, 16 rxns
SDP
Supplier:  Active Motif 53160
Encompass
Cleavage Under Targets and Tagmentation (CUT&Tag) is a method to investigate genomic localization of histone modifications and some transcription factors that reveals interactions between proteins and DNA or identifies DNA binding sites for proteins of interest.
Catalog No. NC1931391