Learn More
Aviva Systems Biology Corp CUTA Antibody - middle region (ARP44681_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CUTA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CUTA. Alternate names: ACHAP, C6orf82, MGC111154ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53977