Learn More
Selleck Chemical LLC Cutamesine Dihydrochloride S2917-5mg
SDP
Catalog No. 503111823
Encompass_Preferred
Cutamesine dihydrochloride (SA4503 dihydrochloride AGY94806 dihydrochloride) is a potent Sigma-1 receptor agonist with an IC50 of 17 4 nM in guinea pig brain membranes
Catalog No. 50-311-1823 Supplier Selleck Chemical LLC Supplier No. S29175MG
Cutamesine dihydrochloride (SA4503 dihydrochloride AGY94806 dihydrochloride) is a potent Sigma-1 receptor agonist with an IC50 of 17 4 nM in guinea pig brain membranes