Learn More
EPICYPHER INC CUTANA ChIC/CUT&RUN Kit, 48 rxns
SDP
Supplier:  EPICYPHER INC 141048
Encompass
The CUTANA ChIC/CUT&RUN Kit contains sufficient materials for 48 CUT&RUN samples and is designed for multi-channel sample pipetting in order to realize the increased experimental throughput advantage of CUT&RUN.
Catalog No. NC1858333