Learn More
EPICYPHER INC CUTANA™ Concanavalin A Conjugated Paramagnetic Beads | 50uL - 50rxns
SDP
Catalog No. NC1831103
Encompass
This product contains Concanavalin A-conjugated to paramagnetic microspheres. Concanavalin A (ConA) is a lectin (carbohydrate-binding protein) that binds specifically to mannosyl- and glucosyl-containing extracellular glycoproteins. The ConA magnetic beads are therefore useful to immobilize cells or nuclei presenting these glycans in their extracellular matrices.
Catalog No. NC1831103 Supplier EPICYPHER INC Supplier No. 211401
This product contains Concanavalin A-conjugated to paramagnetic microspheres. Concanavalin A (ConA) is a lectin (carbohydrate-binding protein) that binds specifically to mannosyl- and glucosyl-containing extracellular glycoproteins. The ConA magnetic beads are therefore useful to immobilize cells or nuclei presenting these glycans in their extracellular matrices.