Learn More
EPICYPHER INC CUTANA™ DNA Purification Beads, 5ml
SDP
Catalog No. NC3822379
Encompass
CUTANA™ DNA Purification Beads harness Solid-Phase Reversible Immobilization (SPRI*) technology to deliver fast, high-yield DNA cleanup and precise size selection for NGS library preparation and other genomic workflows. Optimized PEG and salt concentrations drive efficient, reproducible binding of DNA to paramagnetic beads, ensuring exceptional recovery with minimal bias or carry-over contamination. The formulation matches the bead-to-sample ratio of popular alternatives such as AMPure XP* and SPRIselect*, ensuring CUTANA™ DNA Purification Beads can serve as a drop-in replacement into manual or automated cleanup workflows without any protocol re-optimization.
Catalog No. NC3822379 Supplier EPICYPHER INC Supplier No. 2114075ML
CUTANA™ DNA Purification Beads harness Solid-Phase Reversible Immobilization (SPRI*) technology to deliver fast, high-yield DNA cleanup and precise size selection for NGS library preparation and other genomic workflows. Optimized PEG and salt concentrations drive efficient, reproducible binding of DNA to paramagnetic beads, ensuring exceptional recovery with minimal bias or carry-over contamination. The formulation matches the bead-to-sample ratio of popular alternatives such as AMPure XP* and SPRIselect*, ensuring CUTANA™ DNA Purification Beads can serve as a drop-in replacement into manual or automated cleanup workflows without any protocol re-optimization.