Learn More
EPICYPHER INC CUTANA E. coli Spike-in DNA, 100 ng
SDP
Catalog No. NC1860766
Encompass
Fragmented DNA derived from Escherichia coli (E. coli) can be used as a spike-in control for experimental normalization in Cleavage Under Targets and Release Using Nuclease (CUT&RUN). CUTANA E. coli Spike-in DNA contains sufficient material for 100-200 CUT&RUN samples (high and low abundance targets, respectively).
Catalog No. NC1860766 Supplier EPICYPHER INC Supplier No. 181401
Fragmented DNA derived from Escherichia coli (E. coli) can be used as a spike-in control for experimental normalization in Cleavage Under Targets and Release Using Nuclease (CUT&RUN). CUTANA E. coli Spike-in DNA contains sufficient material for 100-200 CUT&RUN samples (high and low abundance targets, respectively).