Learn More
EPICYPHER INC CUTANA pAG-Tn5 for ChIC/CUT&Tag, 50 rxns
SDP
Catalog No. NC1831101
Encompass
Recombinantly produced in E. coli, CUTANA pAG-Tn5 for ChIC/CUT&Tag is a fusion of Proteins A and G to Transposase Tn5. This construct is useful in performing Cleavage Under Targets and Tagmentation (CUT&Tag). The active dimer of Transposase Tn5 is charged with Illumina adapters and ready to be used immediately in CUT&Tag. CUTANA pAG-Tn5 does not contain an epitope tag, which makes it compatible with tag-mediated CUT&Tag.
Catalog No. NC1831101 Supplier EPICYPHER INC Supplier No. 151017
Recombinantly produced in E. coli, CUTANA pAG-Tn5 for ChIC/CUT&Tag is a fusion of Proteins A and G to Transposase Tn5. This construct is useful in performing Cleavage Under Targets and Tagmentation (CUT&Tag). The active dimer of Transposase Tn5 is charged with Illumina adapters and ready to be used immediately in CUT&Tag. CUTANA pAG-Tn5 does not contain an epitope tag, which makes it compatible with tag-mediated CUT&Tag.