Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Cut Level A3 Knit Sleeve
SDP

Catalog No. 17355202 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-202 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-355-202 Supplier National Safety Apparel™ Supplier No. S00KN18
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Knit sleeve provides cut, puncture, and abrasion protection.

  • Stretches for comfortable fit and full range of motion
  • Worn to elbow or middle of upper arm
  • Provides cut, puncture, and abrasion protection

Specifications

Product Type Cut-resistant Sleeve
Construction 2-Ply
Size Universal
Material Aramid Fiber
Color Yellow
Sterility Non-sterile
Style Standard Sleeve
Disposable Reusable
Closure Type Elastic Ends
Seam Type Reinforced
For Use With (Application) Glass Manufacturing
Certifications/Compliance ANSI/ISEA 105 Cut Level A3, ANSI/ISEA 105 Abrasion Level 2, ANSI/ISEA 105 Puncture Level 5, ANSI/ISEA 105 Heat Level 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.