Learn More
ATCC Cutibacterium acnes
Supplier:  ATCC 29399
Encompass
Cutibacterium acnes strain 3 was isolated from the human forehead. This bacterium is propagated anaerobically and can be used as a bacteriophage host.
Catalog No. 50-238-5046