Learn More
Sino Biological Recombinant Human B7-1/CD80 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10698H02H1
Encompass_Preferred
A DNA sequence encoding the human CD80 (NP_005182.1) (Met1-Asn242) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6548