Learn More
Med Vet International Cuttable Bone Plate, J0374A, 25 Holes, 2.7mm, Each
Catalog No. 5000582708
Encompass_Preferred
Can customize for each surgery. Easy to contour for reconstruction of non loadbearing bone.
Catalog No. 50-005-82708 Supplier Med Vet International Supplier No. JOR374A
Can customize for each surgery. Easy to contour for reconstruction of non loadbearing bone.