Learn More
Med Vet International Cuttable Plate, J0374F, 30 Holes, 2.0mm, Each
Catalog No. 5000582709
Encompass_Preferred
Cuttable Bone Plates, 3.5mm and 2.7mm 2.0mm holes, 5mm spacing, 30 holes
Catalog No. 50-005-82709 Supplier Med Vet International Supplier No. JOR374F
Cuttable Bone Plates, 3.5mm and 2.7mm 2.0mm holes, 5mm spacing, 30 holes