Learn More
Med Vet International Cuttable Plate, J0374G, 25 Holes, 2.7mm, Each
Catalog No. 5000582747
Encompass_Preferred
Cuttable Bone Plates, 3.5mm and 2.7mm 2.7mm holes, 6mm spacing, 25 holes
Catalog No. 50-005-82747 Supplier Med Vet International Supplier No. JOR374G
Cuttable Bone Plates, 3.5mm and 2.7mm 2.7mm holes, 6mm spacing, 25 holes