Learn More
Analytical Sales & Services Cutter for 4mm DBS Punch
Catalog No. 5000804286
Encompass
Replacement Cutter for 4mm DBS Punch (cannot be interchanged between Punch Pod sizes)
Catalog No. 50-008-04286 Supplier Analytical Sales & Services Supplier No. 327904
Replacement Cutter for 4mm DBS Punch (cannot be interchanged between Punch Pod sizes)