Learn More
Analytical Sales & Services Cutter for 5mm DBS Punch
Catalog No. 5000806894
Encompass
Replacement Cutter for 5mm DBS Punch (cannot be interchanged between Punch Pod sizes)
Catalog No. 50-008-06894 Supplier Analytical Sales & Services Supplier No. 327905
Replacement Cutter for 5mm DBS Punch (cannot be interchanged between Punch Pod sizes)