Learn More
Chemglass Life Sciences Cutter, Glass, Diamond Tipped with Turet Head
SDP
Supplier:  Chemglass Life Sciences CG117920
Encompass_Preferred
Cutter, Glass, Diamond Tipped with Turet Head. Nickel plated brass cutter has a diamond tip with a red wooden handle. CG-1179-20 rack has four slots that will accommodate plates from 1/16” to 3/16” (1.5 to 4.75mm) thick.
Catalog No. 50-872-005