Learn More
Microsolv Technology Corporation Cutting Blade, Replacement for Clean Cut90 Tubing Cutter. 1 EA.
Supplier:  Microsolv Technology Corporation 46040R
Encompass_Preferred
This blade is a replacement for the Clean Cut90™ Cutting Tool and has a "flat side" that produces a 90⁰ cut on PEEK or Polymer Tubing.
Catalog No. 50-227-1441