Learn More
STERILAB INC CUTTING BOARDS LABBOARDS PK25
Catalog No. 501317561
Encompass_Preferred
AMNION AND TISSUE processing boards 12"x18" ALL-POLY LABboards designed specifically for processing larger tissue specimens for allografts. Printed with metric perimeter rulers and centimeter field grid. 25-count pack. Disposable.
Catalog No. 50-131-7561 Supplier STERILAB INC Supplier No. 1218AM
AMNION AND TISSUE processing boards 12"x18" ALL-POLY LABboards designed specifically for processing larger tissue specimens for allografts. Printed with metric perimeter rulers and centimeter field grid. 25-count pack. Disposable.