Learn More
Electron Microscopy Sciences Self Healing Cutting Mat 2.5x3"
SDP
Catalog No. 5030946
Encompass_Preferred
These cutting mats are inert, self-healing and can be sterilized with ethanol.
Catalog No. 50-309-46 Supplier Electron Microscopy Sciences Supplier No. 6903723
These cutting mats are inert, self-healing and can be sterilized with ethanol.