Learn More
Electron Microscopy Sciences Self Healing Cutting Mat 6x8"
SDP
Catalog No. 5030944
Encompass_Preferred
These cutting mats are inert, self-healing and can be sterilized with ethanol.
Catalog No. 50-309-44 Supplier Electron Microscopy Sciences Supplier No. 69037
These cutting mats are inert, self-healing and can be sterilized with ethanol.