Learn More
World Precision Instrument Cutting Mats for Biopsy Punches-504620
SDP
Catalog No. NC2100578
Encompass
Use our cutting mats for clean cuts through your tissue samples. Our mats ensure that your cored sample comes out whole without burrs or rough edges. Our 1.5mm thick, five layer composite cutting mats are reversible, inert and self-healing. These mats are designed for use with your  biopsy punches .
This item is not returnable. View return policy
Catalog No. NC2100578 Supplier World Precision Instrument Supplier No. 504620
Use our cutting mats for clean cuts through your tissue samples. Our mats ensure that your cored sample comes out whole without burrs or rough edges. Our 1.5mm thick, five layer composite cutting mats are reversible, inert and self-healing. These mats are designed for use with your  biopsy punches .