Learn More
Ampco Cutting Pliers
SDP
Catalog No. 19173542 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
9 -7/8 in.
Catalog No. Length (English)
19-173-542 9 -7/8 in.
1 options
Catalog No. 19-173-542 Supplier Ampco Supplier No. 41242
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant cutting pliers. Made in the USA with German components.
  • Length: 9 and 7/8 in.
  • Weight: 1.15 lb.
Specifications
Product Type Cutting Pliers
Length (English) 9 -7/8 in.
No. per Pack 1 Ea.