Learn More
Electron Microscopy Sciences EMS High Precision Cutting Tweezer Style 15AGW, 115 mm
SDP
Catalog No. 501930590
Encompass_Preferred
For cutting hair sprigs, electronic component wires, copper, gold, silver and other soft wires. Large head, oblique strong cut at the cutter's tip. Cutting edge is limited to the tip area. 111 mm long. M = 11 mm, N = 3 mm, P = 24 mm.
OAL: 115 mm
Catalog No. 50-193-0590 Supplier Electron Microscopy Sciences Supplier No. 7832504
For cutting hair sprigs, electronic component wires, copper, gold, silver and other soft wires. Large head, oblique strong cut at the cutter's tip. Cutting edge is limited to the tip area. 111 mm long. M = 11 mm, N = 3 mm, P = 24 mm.
OAL: 115 mm