Learn More
Foxx Life Sciences Borosil Reusable Heavy Duty Culture Tubes with PTFE Lined PP Screw Caps and Round Bottoms, 30mL, 100/CS
Catalog No. 501542914
Encompass_Preferred
Borosil Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, Borosil Culture Tubes are easy to handle & seal. Round bottoms & ultra-high clarity. Ideal replacement for media & culture bottles. Borosil Heavy Duty Culture Tubes with Round Bottoms are Avail. from 5mL to 150mL. Also Avail. in Light-Blocking Amber (Product Family 9901). In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
This item is not returnable. View return policy
Catalog No. 50-154-2914 Supplier Foxx Life Sciences Supplier No. 9900010
Borosil Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, Borosil Culture Tubes are easy to handle & seal. Round bottoms & ultra-high clarity. Ideal replacement for media & culture bottles. Borosil Heavy Duty Culture Tubes with Round Bottoms are Avail. from 5mL to 150mL. Also Avail. in Light-Blocking Amber (Product Family 9901). In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.