Learn More
Foxx Life Sciences Borosil Reusable Heavy Duty Culture Tubes with PTFE Lined PP Screw Caps and Flat Bottoms, 30mL, 100/CS
Catalog No. 501542920
Encompass_Preferred
Borosil Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, tubes are easy to seal. Flat bottoms and ultra-high clarity. Ideal replacement for Biju/Bijou, McCartney, and culture or media bottles. Borosil Heavy Duty Culture Tubes with Flat Bottoms are Avail. from 5mL to 50mL. Also Avail. in Light-Blocking Amber (Product Family 9911). In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-154-2920 Supplier Foxx Life Sciences Supplier No. 9910010
Borosil Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, tubes are easy to seal. Flat bottoms and ultra-high clarity. Ideal replacement for Biju/Bijou, McCartney, and culture or media bottles. Borosil Heavy Duty Culture Tubes with Flat Bottoms are Avail. from 5mL to 50mL. Also Avail. in Light-Blocking Amber (Product Family 9911). In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.